EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Development of 6-substituted indolylquinolinones as potent Chek1 kinase inhibitors.
PMID 16990002
Abstract
Through a comparison of X-ray co-crystallographic data for 1 and 2 in the Chek1 active site, it was hypothesized that the affinity of the indolylquinolinone series (2) for Chek1 kinase would be improved via C6 substitution into the hydrophobic region I (HI) pocket. An efficient route to 6-bromo-3-indolyl-quinolinone (9) was developed, and this series was rapidly optimized for potency by modification at C6. A general trend was observed among these low nanomolar Chek1 inhibitors that compounds with multiple basic amines, or elevated polar surface area (PSA) exhibited poor cell potency. Minimization of these parameters (basic amines, PSA) resulted in Chek1 inhibitors with improved cell potency, and preliminary pharmacokinetic data are presented for several of these compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
523267
4-Bromo-2-iodoaniline, 97%
C6H5BrIN