EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Pharmacological correction of energy metabolism disturbance in the inflammatory-dystrophic process in the periodontium].
PMID 16995440
Abstract
The influence of lipoic acid on the energy homeostasis in rats with chronic generalized parodontitis has been investigated. Lipoic acid possesses the energy-saving activity, which is related to an increase in oxidative phosphorylation efficiency.
Related Materials