EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
In vitro study of polyoxyethylene alkyl ether niosomes for delivery of insulin.
PMID 16997517
Abstract
In this study, niosomes of polyoxyethylene alkyl ethers (Brij) were prepared for encapsulation of insulin by film hydration method. Without cholesterol, brij 35 and brij 58 did not form niosomes, apparently because of relatively large polar head groups in comparison with their alkyl chains. The size of vesicles depended on the cholesterol content, charge incorporation or hydrophilicity of surfactants. Entrapment of insulin in bilayer structure of niosomes protected it against proteolytic activity of alpha-chymotrypsin, trypsin and pepsin in vitro. The maximum protection activity was seen in brij 92/cholesterol (7:3 molar ratios) in which only 26.3+/-3.98% of entrapped insulin was released during 24h in simulated intestinal fluid (SIF). The kinetic of drug release for most formulations could be best described by Baker and Lonsdale equation indicating diffusion based delivery mechanism. These results indicate that niosomes could be developed as sustained release oral dosage forms for delivery of peptides and proteins such as insulin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2631
Dihexadecyl phosphate
C32H67O4P