EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Improvement in rod and cone function in mouse model of Fundus albipunctatus after pharmacologic treatment with 9-cis-retinal.
PMID 17003450
Abstract
To assess changes in rod and cone visual functions in a mouse model of Fundus albipunctatus with disrupted 11-cis-retinol dehydrogenase (RDH) genes after pharmacologic treatment with an artificial retinal chromophore. Retinoid levels and photoreceptor functions of Rdh5-/-Rdh11-/- mice at a variety of light intensities were analyzed with normal-phase HPLC and ERG techniques. Production of 11-cis-retinal, the visual pigment chromophore, was suppressed with a potent inhibitor of the retinoid cycle, all-trans-retinylamine (Ret-NH2). The chromophore was replaced by a functional geometric isomer, 9-cis-retinal, delivered by oral gavage. Aberrant cone responses were detected in 12-month-old Rdh5-/-Rdh11-/- mice raised in a 12-hour light/12-hour dark cycle. This cone defect was exacerbated in conditions of low levels of 11-cis-retinal. Administration of 9-cis-retinal increased the rate of dark adaptation and improved cone function in Rdh5-/-Rdh11-/- mice. Disruption of 11-cis-RDHs causes a slowly developing cone dystrophy caused by inefficient cone pigment regeneration. Rod and cone visual function improved significantly in the mouse model of F. albipunctatus after treatment with 9-cis-retinal, suggesting a potential approach to slow the progression of cone dystrophy in affected humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5754
9-cis-Retinal, vitamin A analog
C20H28O