EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer's disease : JAD
Aluminum and Alzheimer's disease, a personal perspective after 25 years.
PMID 17004365
Abstract
It is now 25 years since the publication of our original paper investigating the association aluminum with Alzheimer's disease. This publication reported on the results of scanning electron microscopy coupled x-ray spectrometry microprobe elemental studies of both neurofibrillary tangle-bearing and tangle-free neurons in the hippocampus of cases of Alzheimer's disease and controls. Peaks related to the presence of aluminum were consistently detected within the tangle-bearing neurons. This paper supported the association of aluminum and Alzheimer's disease on the cellular level of resolution and caused considerable interest and discussion. Subsequent work demonstrated prominent evidence of aluminum accumulation in the tangle-bearing neurons of cases of amyotrophic lateral sclerosis/parkinsonism-dementia complex of Guam. This latter observation has now been replicated using five different forms of microanalysis. Finally, using laser microprobe mass analysis, we demonstrated that the abnormally high aluminum-related signal which we originally detected was actually located within the neurofibrillary tangle, itself, and was accompanied by excess concentrations of iron. Although it is unlikely that aluminum represents an etiologic cause of Alzheimer's disease, we believe that this highly reactive element, known to cross-link hyperphosphorylated proteins, may play an active role in the pathogenesis of critical neuropathologic lesion in Alzheimer's disease and other related disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266523
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
11008
Aluminum, grit, ≥97.0% (complexometric)
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
11009
Aluminum, powder, ≥91% (complexometric)
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
310360
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
11010
Aluminum, powder (fine), ≥93% (complexometric)
Al
IRMM523A
Aluminum, IRMM® certified Reference Material, 0.1 mm foil
Al
IRMM523B
Aluminum, IRMM® certified Reference Material, 1.0 mm foil
Al
IRMM523C
Aluminum, IRMM® certified Reference Material, 1.0 mm wire
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al
749036
Aluminum, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.9995% trace metals basis
Al
773964
Aluminum, ingot, L × W 10 mm × 7 mm, 99.99995% trace metals basis
Al