EMAIL THIS PAGE TO A FRIEND
Annals of the Academy of Medicine, Singapore
A report of two families with sarcosinaemia in Hong Kong and revisiting the pathogenetic potential of hypersarcosinaemia.
PMID 17006587
Abstract
Sarcosinaemia is a rare metabolic disorder which has not been reported in Asia. The urine samples of 2 patients were screened as a routine metabolic screening offered for patients with mental retardation in our hospital. We used gas chromatography-mass spectrometry (GC-MS) which is capable of detecting abnormal pattern in amino acids and organic acids. Plasma sarcosine level was further quantified by GC-MS. The same methods were used in the investigations of asymptomatic family members. Urine examination by GC-MS revealed excessive amount of sarcosine in urine (normally undetectable) and their plasma sarcosine levels were raised. The 2 differential diagnoses of presence of sarcosine in urine--glutaric aciduria type II and folate deficiency--were ruled out by the absence of abnormal organic acids in the initial urine screen and by normal serum folate level respectively. Screening of the 2 families identified excessive sarcosine in urine in 2 siblings, one from each family. However, these 2 siblings of indexed patients thus identified have no neurological or developmental problem. Our finding was consistent with the notion that sarcosinaemia is a benign condition picked up coincidentally during screening for mental retardation.