EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Regulation of alpha-chymotrypsin activity on the surface of substrate-functionalized gold nanoparticles.
PMID 17007412
Abstract
A gold nanoparticle functionalized with substrates for alpha-chymotrypsin was fabricated to afford an enzyme modulator that exhibited enzyme-specific activation coupled with general inhibition of other proteases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4760
N-Succinyl-Ala-Ala-Ala-p-nitroanilide, elastase substrate
C19H25N5O8