EMAIL THIS PAGE TO A FRIEND
International journal of gynaecology and obstetrics: the official organ of the International Federation of Gynaecology and Obstetrics
Medical treatment of vulvar squamous cell hyperplasia.
PMID 17010347
Abstract
To evaluate symptomatic response and recurrence rates of graduated topical fluorinated corticosteroid in patients with vulvar squamous cell hyperplasia. Nine hundred seventy-six patients with biopsy-proven vulvar squamous cell hyperplasia from 1990 to 2003 were reviewed in this retrospective study. All patients were treated with graduated topical fluorinated corticosteroid. Data were obtained from hospital records. Symptomatic remission and recurrence rates were noted following six months local therapy. The mean age was 42.55+/-10.93 (15-85). The remission rate was 93.8% in six months. The remission rate was non-significantly higher in postmenopausal patients than that in their premenopausal counterpart (94.9% vs 93.0%, p=0.15). The disease recurred in 6.9% of patients. Of the patients that suffered recurrence 47.5% had persistent disease initially. The patients with following factors older ages (>40 years), postmenopausal period had significantly higher recurrence rates. Four patients with recurrent disease and six patients with persistent disease in the form of vulvar intraepithelial neoplasia I-II or atypical squamous hyperplasia, were treated with skinning vulvectomy. Corticosteroid in the treatment of vulvar squamous cell hyperplasia yielded excellent response rates. In the evaluation of patients without symptomatic relief, the first step should be a vulvar biopsy to exclude the presence of atypical components.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9507
Flumethasone
C22H28F2O5