EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Evaluation of solid adsorbent materials for cryogen-free trapping-gas chromatographic analysis of atmospheric C2-C6 non-methane hydrocarbons.
PMID 17010353
Abstract
Nine commercial solid adsorbent materials (in order of decreasing surface area: Carboxen 1000, Carbosieve S III, molecular sieve 5A, molecular sieve 4A, silica gel, Carboxen 563, activated alumina, Carbotrap and Carboxen 1016) were investigated for their ability to trap and release C2-C6 non-methane hydrocarbons (NMHCs) in atmospheric samples for subsequent thermal desorption gas chromatography-flame ionization detection analysis (GC-FID). Recovery rates for 23 NMHCs and methyl chloride (CH3Cl) were determined. A microtrap filled with the three adsorbents Carbosieve S III, Carboxen 563 and Carboxen 1016 was found to allow for the analysis of the widest range of target analytes. A detection limit of approximately 3pptC [parts per trillion (carbon)] in a 1l air sample and a linear response over a wide range of volatilities and sample volumes was determined for this configuration. Water vapor in the sample air was found to causes interference in trapping and subsequent chromatographic analysis of light NMHCs. A Peltier-cooled, regenerable water trap inserted into the sample flow path was found to mitigate these problems and to allow quantitative and reproducible results for all analytes at all tested humidity conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10197 Carbosieve® Adsorbent, matrix Carbosieve® G, 45-60 mesh, bottle of 5 g
10198 Carbosieve® Adsorbent, matrix Carbosieve® G, 60-80 mesh, bottle of 5 g
10199 Carbosieve® Adsorbent, matrix Carbosieve® G, 80-100 mesh, bottle of 5 g
10189 Carbosieve® Adsorbent, matrix Carbosieve® S-II, 60-80 mesh, bottle of 10 g
10190-U Carbosieve® Adsorbent, matrix Carbosieve® S-II, 80-100 mesh, bottle of 10 g
10184 Carbosieve® Adsorbent, matrix Carbosieve® S-III, 60-80 mesh, bottle of 10 g
28274-U Carbosieve® SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
28942-U Carbosieve® SIII, stainless steel TD tube, unconditioned
20012-U
Carbosieve® SIII, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
29743-U
Carbosieve® SIII, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
25087
Carbotrap® 100, glass TD tube, preconditioned
20242 Carbotrap® 200 Dynatherm 890, 850, for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16
20361 Carbotrap® 201, for use with Dynatherm 850/890 TDU, I.D. × O.D. × L 1 mm × 6 mm × 4.5 in., glass TD tube (focusing tube)
20865 Carbotrap® 201, for use with Dynatherm ACEM 900/901-FF, I.D. × O.D. × L 1 mm × 6 mm × 7.25 in.
20083-U
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
29745-U
Carbotrap® 202, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20013-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
29531-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
30138-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2, unconditioned, pkg of 10 ea
30137-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
29744-U
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29750-U Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20875
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 4.5 in., preconditioned, pkg of 1 ea
28283-U
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
28273-U
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
20370-U Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
28952-U Carbotrap® 300, glass TD tube, unconditioned
28937-U Carbotrap® 300, stainless steel TD tube, unconditioned
21705-U
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, preconditioned, pkg of 10 ea
29532-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea