EMAIL THIS PAGE TO A FRIEND
Journal of the American Society for Mass Spectrometry
Determination of synthetic ferric chelates used as fertilizers by liquid chromatography-electrospray/mass spectrometry in agricultural matrices.
PMID 17010641
Abstract
A high-performance liquid chromatography-electrospray ionization/mass spectrometry (time of flight) method has been developed for the simultaneous determination of synthetic Fe(III)-chelates used as fertilizers. Analytes included the seven major Fe(III)-chelates used in agriculture, Fe(III)-EDTA, Fe(III)-DTPA, Fe(III)-HEDTA, Fe(III)-CDTA, Fe(III)-o,oEDDHA, Fe(III)-o,pEDDHA, and Fe(III)-EDDHMA, and the method was validated using isotope labeled (57)Fe(III)-chelates as internal standards. Calibration curves had R values in the range 0.9962-0.9997. Limits of detection and quantification were in the ranges 3-164 and 14-945 pmol, respectively. Analyte concentrations could be determined between the limits of quantification and 25 muM (racemic and meso Fe(III)-o,oEDDHA and Fe(III)-EDDHMA) or 50 muM (Fe(III)-EDTA, Fe(III)-HEDTA, Fe(III)-DTPA, Fe(III)-CDTA and Fe(III)-o,pEDDHA). The average intraday repeatability values were approximately 0.5 and 5% for retention time and peak area, respectively, whereas the interday repeatability values were approximately 0.7 and 8% for retention time and peak area, respectively. The method was validated using four different agricultural matrices, including nutrient solution, irrigation water, soil solution, and plant xylem exudates, spiked with Fe(III)-chelate standards and their stable isotope-labeled corresponding chelates. Analyte recoveries found were in the ranges 92-101% (nutrient solution), 89-102% (irrigation water), 82-100% (soil solution), and 70-111% (plant xylem exudates). Recoveries depended on the analyte, with Fe(III)-EDTA and Fe(III)-DTPA showing the lowest recoveries (average values of 87 and 88%, respectively, for all agricultural matrices used), whereas for other analytes recoveries were between 91 and 101%. The method was also used to determine the real concentrations of Fe(III)-chelates in commercial fertilizers. Furthermore, the method is also capable of resolving two more synthetic Fe(III)-chelates, Fe(III)-EDDHSA and Fe(III)-EDDCHA, whose exact quantification is not currently possible because of lack of commercial standards.