EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Heart and Circulatory Physiology
TRPC3 channels colocalize with Na+/Ca2+ exchanger and Na+ pump in axial component of transverse-axial tubular system of rat ventricle.
PMID 17012351