EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Facile aerobic photo-oxidation of aldehydes in the presence of catalytic lithium bromide.
PMID 17015991
Abstract
Aldehydes were found to be oxidized to the corresponding carboxylic acid in the presence of catalytic lithium bromide under photo-irradiation.