EMAIL THIS PAGE TO A FRIEND
IDrugs : the investigational drugs journal
Industrial microbiology and biotechnology - 2006 Annual Meeting.
PMID 17016773
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0654
Reveromycin A, solid
C36H52O11