EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Chitosan graft copolymer nanoparticles for oral protein drug delivery: preparation and characterization.
PMID 17025345
Abstract
Several novel functionalized graft copolymer nanoparticles consisting of chitosan (CS) and the monomer methyl methacrylate (MMA), N-dimethylaminoethyl methacrylate hydrochloride (DMAEMC), and N-trimethylaminoethyl methacrylate chloride (TMAEMC), which show a higher solubility than chitosan in a broader pH range, have been prepared by free radical polymerization. The nanoparticles were characterized in terms of particle size, zeta potential, TEM, and FT-IR. These nanoparticles were 150-280 nm in size and carried obvious positive surface charges. Protein-loaded nanoparticles were prepared, and their maximal encapsulation efficiency was up to 100%. In vitro release showed that these nanoparticles provided an initial burst release followed by a slowly sustained release for more than 24 h. These graft copolymer nanoparticles enhanced the absorption and improved the bioavailability of insulin via the gastrointestinal (GI) tract of normal male Sprague-Dawley (SD) strain rats to a greater extent than that of the phosphate buffer solution (PBS) of insulin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
408980
2-(Diethylamino)ethyl methacrylate, contains 1500 ppm MEHQ as inhibitor, 99%
C10H19NO2