EMAIL THIS PAGE TO A FRIEND
Chemistry and physics of lipids
Phase behaviour of stearic acid-stearonitrile mixtures. A thermodynamic study in bulk and at the air-water interface.
PMID 17027948
Abstract
The solid-liquid phase behaviour of stearic acid (SA) and stearonitrile (SN) in binary mixtures was investigated by differential scanning calorimetry (DSC), and the formation of SA-SN mixed monolayers at the air-water interface was followed by surface pressure-area (pi-A) measurements and by Brewster angle microscope (BAM) observation. The solid-liquid phase diagram is a eutectic type phase diagram, with the eutectic composition 0.90
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122580
Stearonitrile, technical grade, ≥90%
C18H35N