EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Alternative water management for controlling simetryn and thiobencarb runoff from paddy fields.
PMID 17033864
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45660
Simetryn, PESTANAL®, analytical standard
C8H15N5S
45687
Thiobencarb, PESTANAL®, analytical standard
C12H16ClNOS