EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Chemical constituents from the south China sea gorgonian coral Subergorgia reticulata].
PMID 17039875
Abstract
Nine compounds, cholesterol (1), ergostra-7,22-diene-3beta, 5alpha, 6beta-triol (2), cholesta-7,22-diene-3beta, 5alpha, 6beta-triol (3),5,8-epidioxycampesta-6,22-dien-3-o1 (4), batyl alcohol (5), theine (6), thymine (7), uracil (8), guanine (9), were isolated from the South China Sea gorgonian coral Subergorgia reticulata and their structures elucidated on the basis of spectral data. All of these compounds were isolated for the first time from this gorgonian coral.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B402
Batyl alcohol, 99%
C21H44O3