EMAIL THIS PAGE TO A FRIEND
Organic letters
Coplanarity in the backbones of ladder-type oligo(p-phenylene) homologues and derivatives.
PMID 17048835
Abstract
p-Tolyl-substituted ladder-type oligo(p-phenylene)s containing three, four, and five phenylene rings were readily synthesized. The uniform aryl substitution of these systems allowed us to determine the coplanarity of the pi-conjugated backbones crystallographically. The intramolecular annulations eliminate almost all of the conformational disorder and enhance the degree of pi-conjugation of the backbones, resulting in significant red shifts in the absorption and emission maxima and lower oxidation potentials in the higher homologues. [structure: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
417149
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 98%
C9H19BO3