EMAIL THIS PAGE TO A FRIEND
Organic letters
Simplification of the Mitsunobu reaction. Di-p-chlorobenzyl azodicarboxylate: a new azodicarboxylate.
PMID 17048845
Abstract
Di-p-chlorobenzyl azodicarboxylate (DCAD) is introduced as a novel, stable, solid alternative to DEAD and DIAD for a variety of Mitsunobu couplings. DCAD/Ph(3)P-mediated reactions in CH(2)Cl(2) generate a readily separable hydrazine byproduct. [reaction: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
680850
Di-(4-chlorobenzyl)azodicarboxylate, 97%
C16H12Cl2N2O4