EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Substituent and solvent effects on electronic spectra of some substituted phenoxyacetic acids.
PMID 17049914
Abstract
The effects of substituents and solvents have been studied through the absorption spectra of nearly 19 para- and ortho-substituted phenoxyacetic acids in the range of 200-400 nm. The effects of substituent on the absorption spectra of compounds under present investigation are interpreted by correlation of absorption frequencies with simple and extended Hammett equations. Effect of solvent polarity and hydrogen bonding on the absorption spectra are interpreted by means of Kamlet equation and the results are discussed.
Related Materials