EMAIL THIS PAGE TO A FRIEND
International journal of hematology
Synergistic effect of arsenic trioxide and flt3 inhibition on cells with flt3 internal tandem duplication.
PMID 17050201
Abstract
Flt3 internal tandem duplication (Flt3-ITD) is a prevalent mutation in acute myeloid leukemia (AML). Flt3-ITD constitutively activates various signaling pathways, including a mitogen-activated protein kinase kinase (MEK)/extracellular signal-regulated kinase (ERK) pathway. Arsenic trioxide (ATO) and MEK inhibition were recently reported to interact synergistically to induce apoptosis in AML cells. In this study, we aimed to clarify whether ATO and Flt3 inhibition would be a more specific and efficient therapy for Flt3-ITD cells. We demonstrate that the combination of ATO and an Flt3 inhibitor, AG1296, profoundly inhibits the growth of Flt3-ITD cells and induces their apoptosis. We further revealed that this combined treatment potently inhibits the ERK activity that might be responsible for cell growth. Moreover, using the Chou-Talalay method, we observed a synergistic growth-inhibitory effect for ATO and AG1296 in Flt3-ITD cells (BaF3-Flt3-ITD, MV4-11, and PL-21 cells), but not in Flt3 wild-type cells (RS4-11 and NB4 cells), for almost all dose ranges tested. Our results provide an experimental basis for a specific and efficient therapy for Flt3-ITD cells that involves combined treatment with Flt3 inhibitors and ATO.