EMAIL THIS PAGE TO A FRIEND
Cancer research
A quantitative in vivo mouse model used to assay inhibitors of tumor-induced angiogenesis.
PMID 1705173
Abstract
An in vivo model of tumor-induced angiogenesis was used to monitor two known inhibitors of angiogenesis, protamine sulfate and the steroid tetrahydro S. Tumor cells entrapped in alginate beads were injected s.c. into mice. Blood vessel induction was measured by two quantitative methods: measurement of hemoglobin at the alginate pellet site, and pooling of radiolabeled RBC to the alginate pellet site. The two methods gave parallel results. Tetrahydro S with or without heparin inhibited blood vessel growth by 50%, and protamine sulfate inhibited blood vessel growth by 85%. These results were supported by gross morphology and histological analysis of the alginate pellet site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8754
5β-Pregnane-3α,17α,21-triol-20-one
C21H34O4