EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Study on the chemical constituents of Curcuma phaeocaulis].
PMID 17059004
Abstract
To investigate the chemical constituents of the rhizome of Curcuma phaeocaulis Val. The constituents were isolated by column chromatography and their structures elucidated by chemical properties and spectroscopic analysis. Five compunds have been isolated and identified as isocurcumenol, curcumenol, alpha-spinasterol, curcumin, and beta-sitosterin-3-O-glucoside. alpha-Spinasterol and alpha-stediol were isolated from Curcuma phaeocaulis Val. for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36236
( −)-Curcumol, analytical standard
C15H24O2