EMAIL THIS PAGE TO A FRIEND
Zhonghua yi xue za zhi
[The mechanism of apoptosis of chronic myeloid leukemia cells induced by the novel p210 bcr/abl inhibitor berbamine].
PMID 17064567
Abstract
To investigate the mechanism of apoptosis of chronic myeloid leukemia (CML) cells induced by the novel p210 bcr/abl inhibitor berbamine. Human Ph+ CML leukemia K562 cells, which express endogenous p210 bcr/abl protein, were cultured in RPMI 1640 and treated with berbamine as indicated time and dose. Flow cytometry (FCM) and Annexin-V-Fluos/PI staining kit were used to evaluate the apoptosis of leukemic cells; FCM and cytoperm/cytofix plus Caspase-3-McAb-PE were employed to measure the leukemic cells with activated Caspase-3. Phosphorylation of p210 bcr/abl protein in the leukemic cells were assessed by a combination of immunoprecipitation (IP) with c-abl antibody and Western blotting with p-Tyr (pY99) antibody. The protein levels of p210 bcr/abl, Hsp90 and Hsp70 in the leukemic cells were determined by Western blotting with antibodies to c-abl, Hsp90, and Hsp70 respectively. After treatment with berbamine at 8 microg/ml for 48 h, the percentages of leukemic cells expressing activated caspase-3 and apoptotic cells were 45.69% and 48.43% respectively. IP and WB results showed that berbamine at low concentration markedly inhibited phosphorylation of p210 bcr/abl protein in the leukemia cells, and the amount of phosphorylated p210 bcr/abl in the leukemia cells exposed to berbamine at 8 microg/ml for 6 h were only 8.41% of that of untreated leukemia cells without the protein levels of p210 bcr/abl down-regulated. Significantly, berbamine also down-regulated chaperone Hsp90 protein, and the amount of Hsp90 protein in the leukemia cells treated with berbamine at 8 microg/ml for 48 h accounted for 18.37% of that of the untreated leukemia cells. Berbamine at 8 microg/ml had no obvious effect on chaperone Hsp70 protein expression associated with the resistance of leukemia cells to apoptosis. (1) Berbamine induces caspase-3-mediated apoptosis of Ph+ leukemia cells through inhibiting phosphorylation of p210 bcr/abl protein and down-regulating its chaperone Hsp90 protein. (2) Unlike Hsp90 inhibitor GA that upregulates Hsp70, berbamine has no obvious effect on chaperone Hsp70 protein expression in leukemia cells, suggesting that berbamine may be a novel class of Hsp90 inhibitor, and further study is required.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
547190
Berbamine dihydrochloride
C37H40N2O6 ·2HCl