EMAIL THIS PAGE TO A FRIEND
The Analyst
Quantitative determination of lead in mixtures of lead(II) halides using solid-state 207Pb NMR spectroscopy.
PMID 17066187
Abstract
We demonstrate a multi-spectrum technique for facile, quantitative determination of lead in solid materials using solid-state (207)Pb NMR that avoids the major problem of uniform excitation across a wide spectral range; the method can be employed without chemical separation or other chemical manipulations and without any prior sample preparation, resulting in a non-destructive analysis, and producing results that are in agreement with gravimetric analyses of mixed samples of the lead halides.
Related Materials