EMAIL THIS PAGE TO A FRIEND
European journal of protistology
Morphology and cell division of the oxytrichids Architricha indica nov. gen., nov. sp., and Histriculus histrio (Müller, 1773), Corliss, 1960 (Ciliophora, Hypotrichida).
PMID 17070749
Abstract
The oxytrichid ciliate Architricha indica nov. gen., nov. sp., isolated from the river Yamuna, Delhi, shows a new combination of characters. It possesses a flexible body, 18 frontal-ventral-transverse (FVT) cirri, 3 right and 2 left marginal cirral rows, 6 dorsal bristle rows and 3 caudal cirri (CC). The FVT cirri arise from 6 primordia, which utilize 6 parental cirri in their origin as is typical of Oxytricha species. Multiple marginal rows (MMR) develop through 5 independent marginal primordia arising "within-row", 1 in each parental marginal row. All the 5 marginal rows are thus morphogenetically active. Such a mode of formation of MMR has not been recorded among oxytrichids and has necessitated separation of A. indica at the generic level. Histriculus, on the other hand, has well-known characteristics, viz. rigid body, confluent marginal rows and absence of CC. The morphogenesis of Histriculus histrio has been described by Berger and Foissner [1997. Cladistic relationships and generic characterization of oxytrichid hypotrichs (Protozoa, Ciliophora). Arch. Protistenkd. 148, 125-155]. Reinvestigation of very early stages of development revealed that (i) the FVT cirral primordia utilize kinetosomes from 5 parental FVT cirri, (ii) the primordium II of the proter is of a composite origin: kinetosomes from the oral primordium merge with the primordium II that originates from the buccal cirrus II/2 and (iii) the FVT primordia V and VI for the 2 daughter cells arise sequentially from the parental cirrus V/4. Thus, the genus Histriculus exhibits a new combination of characters with respect to the origin of FVT cirri, an additional pattern to be added to the known 6 patterns of FVT development in oxytrichids [Berger and Foissner, 1997; Berger, H., 1999. Monograph of the Oxytrichidae (Ciliophora, Hypotrichida), Kluwer Academic Publishers, Dordrecht/Boston/London].