EMAIL THIS PAGE TO A FRIEND
Current opinion in infectious diseases
Ivermectin: uses and impact 20 years on.
PMID 17075336
Abstract
Ivermectin was first discovered and used in veterinary medicine over 20 years ago. This review highlights some of the recent published research from 2005 through June 2006 on the use of ivermectin in both helminth and arthropod infection. In recent years, several published studies have detailed the expanding role for ivermectin in multiple endo and ectoparasitic infections, including scabies, pediculosis, soil transmitted helminths, gnathostomiasis and myiasis. In addition, there is increasing experience with parenteral ivermectin for the treatment of disseminated strongyloidiasis. The success of ivermectin in reducing Onchocerca volvulus and Wuchereria bancrofti transmission through universal treatment in disease control programs continues to be well documented, but recent epidemiologic data describe suboptimal response to ivermectin by O. volvulus in a minority of individuals, the molecular markers for which are currently under investigation. Over 20 years of research and clinical use have advanced ivermectin from its beginnings as a veterinary anthelmintic to its significant role in several successful disease control programs. Nevertheless, further research is needed to understand the basis for suboptimal response and to better define optimal drug regimens for varying diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8898
Ivermectin