EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Effects of isopropyl palmitate on the skin permeation of drugs.
PMID 17077540
Abstract
The model penetrants oxaprozin, nimesulide, gliclazide, and ribavirin, because of their different lipophilicities, were selected to assess the enhancing activity of pre-treatment solutions consisting of isopropyl palmitate (IP) in ethanol (5%, 10%, 15%and 20%, w/w, respectively) across excised rat skin using Franz diffusion cells and HPLC detection. All pre-treatment solutions produced a significant increase in the flux and permeation of all four penetrants (p<0.001) and a relationship between penetrant lipophilicity and enhancement effect was observed. The general order of IP effectiveness at concentration was 20%>15%>10%>5% (w/w). The lag-time of drugs did not significantly change except for ribavirin.