EMAIL THIS PAGE TO A FRIEND
Organic letters
Fully automated continuous flow synthesis of 4,5-disubstituted oxazoles.
PMID 17078685
Abstract
[Structure: see text] A multipurpose mesofluidic flow reactor capable of producing gram quantities of material has been developed as an automated synthesis platform for the rapid on-demand synthesis of key building blocks and small exploratory libraries. The reactor is configured to provide the maximum flexibility for screening of reaction parameters that incorporate on-chip mixing and columns of solid supported reagents to expedite the chemical syntheses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226319
Ethyl isocyanoacetate, 95%
C5H7NO2