EMAIL THIS PAGE TO A FRIEND
American journal of veterinary research
In vitro effects of hydrochloric acid and various concentrations of acetic, propionic, butyric, or valeric acids on bioelectric properties of equine gastric squamous mucosa.
PMID 17078749
Abstract
To compare the effects of hydrochloric acid (HCl) and various concentrations of volatile fatty acids (VFAs) on tissue bioelectric properties of equine stomach nonglandular (NG) mucosa. Gastric tissues obtained from 48 adult horses. NG gastric mucosa was studied by use of Ussing chambers. Short-circuit current (Isc) and potential difference (PD) were measured and electrical resistance (R) and conductance calculated for tissues after addition of HCl and VFAs (5, 10, 20, and 40 mM) in normal Ringer's solution (NRS). Mucosa exposed to HCl in NRS (pH of 1.5 and, to a lesser extent, 4.0) had a significant decrease in Isc, PD, and R, whereas tissues exposed to acetic acid at a pH of < 4.0, propionic and butyric acids at a pH of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75054
Valeric acid, analytical standard
C5H10O2