EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Inhibitory effect of Andrographis paniculata extract and its active diterpenoids on platelet aggregation.
PMID 17081514
Abstract
Andrographis paniculata has been widely used for the prevention and treatment of common cold especially in Asia and Scandinavia. The three active diterpenoids from this plant, including aqueous plant extracts, were investigated for the inhibitory effect on platelet aggregation in vitro. The results indicated that andrographolide (AP(1)) and 14-deoxy-11,12-didehydroandrographolide (AP(3)) significantly inhibited thrombin-induced platelet aggregation in a concentration-(1-100 microM) and time-dependent manner while neoandrographolide (AP(4)) had little or no activity. AP(3) exhibited higher antiplatelet activity than AP(1) with IC(50) values ranging from 10 to 50 microM. The inhibitory mechanism of AP(1) and AP(3) on platelet aggregation was also evaluated and the results indicated that the inhibition of extracellular signal-regulated kinase1/2 (ERK1/2) pathway may contribute to antiplatelet activity of these two compounds. In addition, standardized aqueous extracts of A. paniculata containing different amounts of AP(3) inhibited thrombin-induced aggregation to different degrees. The extracts significantly decreased platelet aggregation in a concentration-(10-100 microg/ml) and time-dependent manner. However, the extract with high level of AP(3) (Extract B) (IC(50) values=50-75 microg/ml) showed less inhibitory activity against thrombin than the extract with lower level of AP(3) (Extract A) (IC(50) values=25-50 microg/ml). These results indicate that the standardized A. paniculata extract may contain other antiplatelet compounds rather than AP(1) and AP(3), which contribute to high antiplatelet activity. Therefore, the consumption of A. paniculata products may help to prevent or treat some cardiovascular disorders i.e. thrombosis; however, it should be used with caution by patients with bleeding disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49879
Neoandrographolide, analytical standard
C26H40O8