EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Determination of chlorpyrifos and its major breakdown products in technical formulations.
PMID 1708301
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45664
Sulfotep, PESTANAL®, analytical standard
C8H20O5P2S2