EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Effects of phthalate esters on the sensitization phase of contact hypersensitivity induced by fluorescein isothiocyanate.
PMID 17083357
Abstract
Many different types of phthalate ester are used as plasticizers and are thus found in the air. There have been several studies that suggest an association between allergies and phthalate esters. We previously found that di-butyl phthalate (DBP) has an adjuvant effect in a mouse contact hypersensitivity model, in which fluorescein isothiocyanate (FITC) is involved as an immunogenic hapten. We examined whether other phthalate esters enhance the process of sensitization to FITC by facilitating the trafficking of FITC-presenting dendritic cells or macrophages from skin sites to draining lymph nodes. Mice were epicutaneously sensitized with FITC dissolved in acetone containing a phthalate ester. Sensitization was evaluated as ear swelling after a challenge with FITC. Draining lymph node cells obtained 24 h after skin sensitization were examined for FITC fluorescence by means of flow cytometry. FITC-positive cells were characterized with anti-CD11c and anti-CD11b by three-colour flow cytometry. When mice were sensitized with FITC in acetone containing DBP or di-n-propyl phthalate (DPP), strong enhancement of the ear-swelling response was observed. Di-methyl phthalate (DMP) and di-ethyl phthalate (DEP) were less effective but produced some enhancement. Consistent enhancement was not observed with di-(2-ethylhexyl) phthalate or di-isononyl phthalate. Upon sensitization in the presence of DBP or DPP, the number of FITC-positive dendritic cells (total CD11c+ as well as CD11c+/CD11b+) was increased in draining lymph nodes. As to the other four phthalate esters, there was no significant increase in the FITC-positive cell number in the draining lymph nodes. During the process of sensitization to FITC, DBP, and DPP exert strong adjuvant effects that are associated with enhancement of trafficking of antigen-presenting dendritic cells from the skin to draining lymph nodes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45624
Dipropyl phthalate, PESTANAL®, analytical standard
C14H18O4
290602
Dipropyl phthalate, 98%
C14H18O4