EMAIL THIS PAGE TO A FRIEND
Brain research
A1 adenosine receptor-mediated modulation of neuronal ATP-sensitive K channels in rat substantia nigra.
PMID 17084818
Abstract
ATP-sensitive K (K(ATP)) channels, widely expressed in cytoplasmic membranes of neurons, couple cell metabolism to excitability. They are considered to play important roles in controlling seizure activity during hypoxia and in neuroprotection against cell damage during hypoxia, ischemia and excitotoxicity. It is known that adenosine augments the opening of cardiac surface K(ATP) channels by reducing the sensitivity of these channels to ATP blockade. We investigated whether a similar modulation occurs in neuronal channels. Whole cell voltage-clamp recordings were made using rat midbrain slices to record the membrane current and conductance in principal neurons of the substantia nigra pars compacta (SNc). When the pipette solution contained 1 mM ATP, the membrane current at -60 mV and cellular conductance remained stable for at least 15 min. When slices were treated with (-)-N(6)-2-phenylisopropyl adenosine (R-PIA), a selective agonist for A(1) adenosine receptors, in the same condition, the outward current developed slowly to the amplitude of 109.9+/-26.6 pA, and conductance increased to 229+/-50% of the baseline. These changes were strongly inhibited by 200 microM tolbutamide, a K(ATP) channel blocker, suggesting that opening of K(ATP) channels mediated these changes. Pretreatment with 8-cyclopentyltheophylline (CPT), a selective A(1) adenosine receptor antagonist, abolished the outward current and conductance increases. Treatment of adenosine resulted in the similar changes sensitive to tolbutamide. These changes were abolished by CPT. These results suggest that activation of A(1) adenosine receptors promotes the opening of K(ATP) channels in principal neurons of the SNc by removing the blockade by ATP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4532
(−)-N6-(2-Phenylisopropyl)adenosine, solid
C19H23N5O4