EMAIL THIS PAGE TO A FRIEND
Izvestiia Akademii nauk. Seriia biologicheskaia
[Variation in sex pheromone composition in large fruit-tree tortrix Archips podana Scop. (Lepidoptera: Tortricidae)].
PMID 17086968
Abstract
Variation in sex pheromone composition was studied in large fruit-tree tortrix. The pheromone contained cis-11-tetradecenyl acetate and trans-11-tetradecenyl acetate as macrocomponents as well as microcomponents cis-11-tetradecenol and trans-11-tetradecenol with the prevalence of cis-11-tetradecenyl acetate and cis-11-tetradecenol. The pheromone composition depended in part on the genotype of the male parent. Inbreeding increased the proportion of trans-11-tetradecenyl acetate and trans-11-tetradecenol.