EMAIL THIS PAGE TO A FRIEND
Danish medical bulletin
Metallothionein I + II expression and roles during neuropathology in the CNS.
PMID 17087875
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A68203
6-Aminonicotinamide, 99%
C6H7N3O