EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Formation and rapid export of the monochlorobimane-glutathione conjugate in cultured rat astrocytes.
PMID 17089195
Abstract
Monochlorobimane (MCB) is often used to visualize glutathione (GSH) levels in cultured cells, since it is quickly converted to a fluorescent GSH conjugate (GS-MCB). To test for consequences of MCB application on the GSH metabolism of astrocytes, we have studied rat astrocyte-rich primary cultures as model system. MCB caused a concentration dependent rapid decrease in the cellular GSH content. Simultaneously, a transient accumulation of GS-MCB in the cells was observed with a maximal content 5 min after MCB application. The cellular accumulation was followed by a rapid release of GS-MCB into the medium with a maximal initial export rate of 27.9 +/- 6.5 nmol h(-1) mg protein(-1). Transporters of the family of multidrug resistance proteins (Mrps) are likely to be involved in this export, since the Mrp inhibitor MK571 lowered the export rate by 60%. These data demonstrate that, due to its rapid export from astrocytes, GS-MCB is only under well-defined conditions a reliable indicator of the cellular GSH concentration and that MK571 can be used to maintain maximal GS-MCB levels in astrocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69899
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)
C10H11ClN2O2