EMAIL THIS PAGE TO A FRIEND
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists
Overview of radiopaque drugs: 1895-1931.
PMID 17090746