EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Hydroaminomethylation with novel rhodium-carbene complexes: an efficient catalytic approach to pharmaceuticals.
PMID 17091522
Abstract
Starting from [{Rh(cod)Cl}(2)] and 1,3-dimesitylimidazole-2-ylidenes the novel [RhCl(cod)(carbene)] complexes 1-5 have been synthesized, characterized, and tested in the hydroaminomethylation of aromatic olefins. The influence of different ligands and reaction parameters on the catalytic activity was investigated in detail applying 1,1-diphenylethylene and piperidine as a model system. The scope and limitations of the novel catalysts is shown in the preparation of 16 biologically active 1-amino-3,3-diarylpropenes. In general, high chemo- and regioselectivity as well as good yields of the desired products were achieved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D206806
1,1-Diphenylethylene, 97%
C14H12