EMAIL THIS PAGE TO A FRIEND
Chemical reviews
Hexafluoroacetone as protecting and activating reagent: new routes to amino, hydroxy, and mercapto acids and their application for peptide and glyco- and depsipeptide modification.
PMID 17091933
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295353
Hexafluoroacetone, 97%
C3F6O
139238
Hexafluoroacetone trihydrate, 98%
C3F6O · 3H2O