EMAIL THIS PAGE TO A FRIEND
Environmental sciences : an international journal of environmental physiology and toxicology
Effect of aspect ratio on chemical reactions on microchip.
PMID 17096000
Abstract
Parallel two-phase laminar flow, which is formed when two solutions flow in microchannels, has been developed and has advanced unique research in the area of microchip analysis. In two-phase laminar flow, channel size has a significant effect on the efficiency of chemical reactions. However, the sizes of microchannels vary greatly in many studies. In this paper, we report on the effect of microchannel size on chemical reactions on a microchip. Aspect ratio is defined as the ratio of depth to width of a microchannel. Five microchips with different aspect ratios (from 0.50 to 2.00) were fabricated by mechanical machining. The reaction of nitrous acid and Saltzman reagent was carried out on these microchips and the absorbance was measured on-line in a capillary tube, which was attached to the outlet on the microchip. The results showed that the color reaction occurred more efficiently as the aspect ratio increased. This result is expected to be useful when determining the size of microchannels.