EMAIL THIS PAGE TO A FRIEND
Journal of minimally invasive gynecology
Danazol treatment of endometriosis-associated pain.
PMID 17097573
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7662
Nafarelin acetate salt hydrate, ≥95% (HPLC), solid
C66H83N17O13 · xC2H4O2 · yH2O