EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of alpha-fluorosulfonate and alpha-fluorosulfonamide analogues of a sulfated carbohydrate.
PMID 17107086
Abstract
The first synthesis of sulfonamide and sulfonate analogues of a sulfated carbohydrate in which the ester oxygen of the sulfate is replaced with a CHF or CF2 group is reported. This was accomplished by electrophilic fluorination of the protected sulfonate and sulfonamide precursors. [reaction: see text].