EMAIL THIS PAGE TO A FRIEND
Lakartidningen
[New agents against atherosclerosis tested. Alarming findings, ACAT inhibitors seem to have proatherogenic effects].
PMID 17117657
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0190
Avasimibe, ≥98% (HPLC)
C29H43NO4S