EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Coiled molecules in spring loaded devices.
PMID 17117825
Abstract
A cylindrical capsule containing a coiled alkane incorporates spacers. The alkane lengthens and gauche strain is relieved. The spacer can be removed to reset the system to the original state. The system represents a spring-loaded device that operates reversibly in response to acids and bases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7305
Glycoluril, 97%
C4H6N4O2