EMAIL THIS PAGE TO A FRIEND
Drugs
Clinical pharmacology of cilazapril.
PMID 1712269
Abstract
In clinical pharmacology studies, cilazapril, after its bioactivation to cilazaprilat, was characterised as a potent, reversible angiotensin converting enzyme (ACE) inhibitor with a terminal half-life of 30 to 50 hours, which is consistent with saturable binding to ACE. Despite the arterial vasodilatation, only slight increases in heart rate occurred during cilazapril administration. Cilazapril had no acute effect on cardiovascular reflexes, and increased effective renal plasma flow slightly. Glomerular filtration rate remained unaltered. A close positive correlation was found between the cilazaprilat plasma concentration and degree of ACE inhibition. The potency of cilazaprit, defined as the concentration of cilazaprilat causing 50% inhibition of ACE, was approximately 1 microgram/L plasma. In short term studies in patients with hypertension, it appeared that more than 90% inhibition of plasma ACE was needed to obtain blood pressure reduction. Results of various dose-response studies established the indirect relationship between dose, the plasma concentration of the drug, and the blood pressure response, and identified the dose producing the maximal effect to be 5mg. Cilazapril inhibited ACE for a relatively long period which was extended in patients with severe chronic renal impairment or hepatic failure. In these patients a reduction of the dose and/or less frequent administration is recommended. There was no clinically relevant interaction of cilazapril with food, furosemide (frusemide), digoxin or coumarins. The effects of hydrochlorothiazide on sodium and chloride excretion were potentiated by cilazapril, and an additive effect of propranolol and nitrendipine on the blood pressure response to cilazapril was observed. An interaction with indomethacin and cilazapril might occur, potentially reducing the blood pressure-lowering effect of cilazapril. In general, cilazapril was well tolerated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2174000 Cilazapril, European Pharmacopoeia (EP) Reference Standard
C22H31N3O5 · H2O