EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Identification and characterization of degradation products of dicloxacillin in bulk drug and pharmaceutical dosage forms.
PMID 17125952
Abstract
Impurity profiling of dicloxacillin sodium bulk drug and pharmaceutical dosage forms subjected to stability studies is evaluated. Of many impurities detected in HPLC analysis, three were not reported in the literature. The impurities have been identified by LC-MS; isolated by preparative HPLC; and characterised by NMR, Mass spectroscopy and IR. Pure impurities obtained by isolation were co-injected with dicloxacillin sodium sample to confirm the retention times in HPLC. Structure elucidation of these degradation products by spectral data has been discussed in detail.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9016
Dicloxacillin sodium salt monohydrate
C19H16Cl2N3NaO5S·H2O