EMAIL THIS PAGE TO A FRIEND
Revue des maladies respiratoires
[Thoracic hydatid cysts: news].
PMID 17127973
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34176
Oxfendazole, VETRANAL, analytical standard
C15H13N3O3S