EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Characterization of a bifunctional aminoacylase/carboxypeptidase from radioresistant bacterium Deinococcus radiodurans R1.
PMID 17129628
Abstract
The gene encoding a Deinococcus radiodurans R1 bifunctional aminoacylase/carboxypeptidase (DR_ACY/CP) was amplified by polymerase chain reaction and cloned into pQE-30 to generate pQE-DRAC. The cloned gene consists of an open reading frame of 1197 bp encoding a protein with a molecular mass of 42,729 Da. The predicted amino acid sequence shows high homology with those of Geobacillus kaustophilus aminoacylase, Geobacillus stearothermophilus aminoacylase, Pyrococcus horikoshii carboxypeptidase/aminoacylase and Thermoanaerobacter tengcongensis aminoacylase/carboxypeptidase. The expressed enzyme was purified from the crude extract of IPTG-induced Escherichia coli M15 (pQE-DRAC) to homogeneity by nickel-chelate chromatography. The molecular mass of the purified enzyme was determined to be 43kDa by SDS-PAGE. Maximal aminoacylase activity with N-acetyl-methionine as the substrate occurred at pH 8.0 and 40 degrees C in the sodium phosphate buffer. The aminoacylase activity was strongly inhibited by metal-chelating agents, and was largely restored by divalent cations, such as Co(2+), Mn(2+) and Ni(2+). The purified enzyme had broad specificity toward N-acetylated L-amino acids as well as N-CBZ-peptides. Carboxypeptidase activity of DR_ACY/CP to N-CBZ-Gly-Ala exhibited K(m) and k(cat) values of 4.3mM and 28s(-1), respectively. The enzyme also had activity toward the cell wall-related substrates, D-Ala-Gly, D-Ala-Gly-Gly and L-Orn-L-Ala.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1141
Z-Phe-Leu
C23H28N2O5