EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
Detection of gunshot residues on cadaveric skin using sodium rhodizonate and a counterstain.
PMID 17129998
Abstract
We report a staining method for cadaveric tissue using sodium rhodizonate as a skin marker for gunshot residues and a counterstain for the surrounding connective tissue. We studied six well preserved subjects who had died of close range gunshot injury. Skin fragments were removed from the bullet entrance hole including both the disrupted area and adjacent macroscopically intact tissue. Because microscopic examination of postmortem material is difficult after histomorphologic alterations already have occurred as a consequence of postmortem tissue changes, it is necessary to use a staining method that, while detecting gunshot residues, can also make skin cell constituents recognizable from both qualitative and quantitative perspectives. Triphenylmethane dyes (acid fuchsin, aniline blue WS, light green SF yellowish, brilliant green and ethyl green) have proven appropriate for the purpose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392340
Potassium rhodizonate, technical grade, 85%
C6K2O6